2-Methyl-6-(trifluoromethyl)nicotinic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF19.2.5 CFF19.5 017016-2.5G 017016-5G
CHMPCFF19.2.5EA 0 GBP
CHMPCFF19.2.5 CHMPCFF19.5 FLUO017016-2.5G FLUO017016-5G
2-Methyl-6-(trifluoromethyl)nicotinic acid
2-Methyl-6-(trifluoromethyl)nicotinic acid
Formula: C₈H₆F₃NO₂
MW: 205.14 g/mol
MDL Number: MFCD01311987
CAS Number: 261635-93-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR