2-Methyl-6-(trifluoromethyl)nicotinic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
005903-5G
005903-1G
FLUO005903-5GEA
0
GBP
FLUO005903-5G
FLUO005903-1G
2-Methyl-6-(trifluoromethyl)nicotinic acid
2-Methyl-6-(trifluoromethyl)nicotinic acid
Formula:
C₈H₆F₃NO₂ MW: 205.14 g/mol |
MDL Number:
MFCD01311987 CAS Number: 261635-93-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...