6-(Trifluoromethyl)nicotinic acid hydrazide

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22568G1 AS22568G5
ACTIAS22568G1EA 0 GBP
ACTIAS22568G1 ACTIAS22568G5
6-(Trifluoromethyl)nicotinic acid hydrazide
6-(Trifluoromethyl)nicotinic acid hydrazide
Formula: C₇H₆F₅N₃O
MDL Number: MFCD01862651
CAS Number: 386715-32-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR