6-(Trifluormethyl)nicotinonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20062G1 AS20062G25 AS20062G5
ACTIAS20062G1EA 0 GBP
ACTIAS20062G1 ACTIAS20062G25 ACTIAS20062G5
6-(Trifluormethyl)nicotinonitrile
6-(Trifluormethyl)nicotinonitrile
Formula: C₇H₃F₃N₂
MDL Number: MFCD03001347
CAS Number: 216431-85-5
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR