3-Amino-2-(4-fluorophenoxy)pyridine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF379.2.5 CFF379.10 CFF379.25 017405-2.5G 017405-10G 017405-25G 017405-1G
CHMPCFF379.2.5EA 0 GBP
CHMPCFF379.2.5 CHMPCFF379.10 CHMPCFF379.25 FLUO017405-2.5G FLUO017405-10G FLUO017405-25G FLUO017405-1G
3-Amino-2-(4-fluorophenoxy)pyridine
3-Amino-2-(4-fluorophenoxy)pyridine
Formula: C₁₁H₉FN₂O
MDL Number: MFCD00067803
CAS Number: 175135-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR