4,4'-(Hexafluoroisopropylidene)diphthalic anhydride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
5,5'-(Perfluoropropane-2,2-diyl)bis(isobenzofuran-1,3-dione), 2,2-Bis(1,3-dihydro-1,3-dioxobenzo[c]furan-5-yl)hexafluoropropane
006565-5G
006565-25G
006565-100G
006565-10G
FLUO006565-5GEA
0
GBP
FLUO006565-5G
FLUO006565-25G
FLUO006565-100G
FLUO006565-10G
4,4'-(Hexafluoroisopropylidene)diphthalic anhydride
4,4'-(Hexafluoroisopropylidene)diphthalic anhydride
Formula:
C₈H₄O₃ |
MDL Number:
MFCD00039143 CAS Number: 1107-00-2 EINECS: 201-607-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...