4'-(2-Fluorophenyl)propiophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-(2'-Fluorobiphenyl-4-yl)propan-1-one

003838-1G 003838-5G
FLUO003838-1GEA 0 GBP
FLUO003838-1G FLUO003838-5G
4'-(2-Fluorophenyl)propiophenone
4'-(2-Fluorophenyl)propiophenone
Formula: C₁₅H₁₃FO
MDL Number: MFCD00236612
CAS Number: 37989-92-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR