4-(4-Fluorobenzyloxy)benzaldehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-((4-Fluorobenzyl)oxy)benzaldehyde

008325-1G 008325-5G
FLUO008325-1GEA 0 GBP
FLUO008325-1G FLUO008325-5G
4-(4-Fluorobenzyloxy)benzaldehyde
4-(4-Fluorobenzyloxy)benzaldehyde
Formula: C₁₄H₁₁FO₂
MDL Number: MFCD00173650
CAS Number: 56442-17-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR