4-Benzyloxy-3-chlorophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Benzyloxy-3-chlorobenzeneboronic acid, (4-(Benzyloxy)-3-chlorophenyl)boronic acid

BB-2036-1G BB-2036-100G BB-2036-5G BB-2036-25G
COBBBB-2036-1GEA 0 GBP
COBBBB-2036-1G COBBBB-2036-100G COBBBB-2036-5G COBBBB-2036-25G
4-Benzyloxy-3-chlorophenylboronic acid
4-Benzyloxy-3-chlorophenylboronic acid
Formula: C₁₃H₁₂BClO₃
MW: 262.50 g/mol
MDL Number: MFCD04115642
CAS Number: 845551-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR