2-Bromo-6-(4-morpholino)pyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Bromo-6-morpholinopyridine

AS22377G1 AS22377G25 AS22377G5
ACTIAS22377G1EA 32 GBP
ACTIAS22377G1 ACTIAS22377G25 ACTIAS22377G5
2-Bromo-6-(4-morpholino)pyridine
2-Bromo-6-(4-morpholino)pyridine
Formula: C₉H₁₁BrN₂O
MW: 243.1004 g/mol
MDL Number: MFCD07772824
CAS Number: 332134-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR