5'-Chloro-2'-methoxy-4'-methylacetophenone
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
019290-5G
019290-25G
FLUO019290-5GEA
0
GBP
FLUO019290-5G
FLUO019290-25G
5'-Chloro-2'-methoxy-4'-methylacetophenone
5'-Chloro-2'-methoxy-4'-methylacetophenone
Formula:
C₁₀H₁₁ClO₂ |
MDL Number:
MFCD03789140 CAS Number: 28478-40-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...