Acetophenone-p-toluenesulphonylhydrazone ≥98%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H53474.06
H53474.14
H53474.06EA
41.6
GBP
H53474.06
H53474.14
Acetophenone-p-toluenesulphonylhydrazone ≥98%
Acetophenone p-toluenesulphonylhydrazone
Purity: 98%
CAS No.: 4545-21-5
Molecular Formula: C15H16N2O2S
Formula Weight: 288.36
Melting Point: 144-149°
MDL No.: MFCD01140191
Formula:
C₁₅H₁₆N₂O₂S Melting Pt: 144…149 °C Storage Temperature: Ambient |
MDL Number:
MFCD01140191 CAS Number: 4545-21-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...