3-Bromo-3',4'-dimethoxybenzophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

022697-1G 022697-5G
FLUO022697-1GEA 0 GBP
FLUO022697-1G FLUO022697-5G
3-Bromo-3',4'-dimethoxybenzophenone
3-Bromo-3',4'-dimethoxybenzophenone
Formula: C₁₅H₁₃BrO₃
MDL Number: MFCD02127297
CAS Number: 681469-97-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR