4'-(4-Bromobenzyloxy)acetophenone
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-(4-((4-Bromobenzyl)oxy)phenyl)ethanone
022112-25G
022112-5G
FLUO022112-25GEA
0
GBP
FLUO022112-25G
FLUO022112-5G
4'-(4-Bromobenzyloxy)acetophenone
4'-(4-Bromobenzyloxy)acetophenone
Formula:
C₁₅H₁₃BrO₂ MW: 305.170 g/mol |
MDL Number:
MFCD03999373 CAS Number: 694443-80-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...