2'-(4-Bromobenzyloxy)acetophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

023240-5G 023240-25G
FLUO023240-5GEA 0 GBP
FLUO023240-5G FLUO023240-25G
2'-(4-Bromobenzyloxy)acetophenone
2'-(4-Bromobenzyloxy)acetophenone
Formula: C₁₅H₁₃BrO₂
MDL Number: MFCD06411652
CAS Number: 845866-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR