3-Butynyl toluene-4-sulphonate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

013129-1G 013129-5G 013129-25G
FLUO013129-1GEA 0 GBP
FLUO013129-1G FLUO013129-5G FLUO013129-25G
3-Butynyl toluene-4-sulphonate
3-Butynyl toluene-4-sulphonate
Formula: C₁₁H₁₂O₃S
MDL Number: MFCD00041687
CAS Number: 23418-85-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR