2-Methoxy-4-methylaniline hydrochloride
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AN-1233-5G
AN-1233-1G
COBBAN-1233-5GEA
0
GBP
COBBAN-1233-5G
COBBAN-1233-1G
2-Methoxy-4-methylaniline hydrochloride
2-Methoxy-4-methylaniline hydrochloride
Formula:
C₈H₁₂ClNO MW: 173.6400 g/mol |
MDL Number:
MFCD09878311 CAS Number: 71288-98-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...