5'-Fluoro-2'-methoxy-3'-nitroacetophenone
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
019297-1G
019297-5G
FLUO019297-1GEA
0
GBP
FLUO019297-1G
FLUO019297-5G
5'-Fluoro-2'-methoxy-3'-nitroacetophenone
5'-Fluoro-2'-methoxy-3'-nitroacetophenone
Formula:
C₉H₈FNO₄ |
MDL Number:
MFCD03789144 CAS Number: 685892-18-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...