4'-(Methylsulphonyl)acetophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

011302-5G 011302-25G
FLUO011302-5GEA 0 GBP
FLUO011302-5G FLUO011302-25G
4'-(Methylsulphonyl)acetophenone
4'-(Methylsulphonyl)acetophenone
Formula: C₉H₁₀O₃S
MDL Number: MFCD00025072
CAS Number: 10297-73-1
EINECS: 233-672-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR