4'-(4-Chlorophenoxy)acetophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

022127-25G 022127-5G
FLUO022127-25GEA 0 GBP
FLUO022127-25G FLUO022127-5G
4'-(4-Chlorophenoxy)acetophenone
4'-(4-Chlorophenoxy)acetophenone
Formula: C₁₄H₁₁ClO₂
MDL Number: MFCD02246925
CAS Number: 41150-48-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR