5'-Chloro-2'-methoxy-3'-nitroacetophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

019301-5G 019301-25G
FLUO019301-5GEA 0 GBP
FLUO019301-5G FLUO019301-25G
5'-Chloro-2'-methoxy-3'-nitroacetophenone
5'-Chloro-2'-methoxy-3'-nitroacetophenone
Formula: C₉H₈ClNO₄
MDL Number: MFCD00191921
CAS Number: 7195-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR