4-Amino-3-chloro-5-methylbenzonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005509-1G 005509-5G
FLUO005509-1GEA 0 GBP
FLUO005509-1G FLUO005509-5G
4-Amino-3-chloro-5-methylbenzonitrile
4-Amino-3-chloro-5-methylbenzonitrile
Formula: C₈H₇ClN₂
MW: 166.61 g/mol
MDL Number: MFCD00041476
CAS Number: 158296-69-6
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR