4'-(2-Bromo-4-fluorophenoxy)acetophenone
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
023247-5G
023247-25G
FLUO023247-5GEA
0
GBP
FLUO023247-5G
FLUO023247-25G
4'-(2-Bromo-4-fluorophenoxy)acetophenone
4'-(2-Bromo-4-fluorophenoxy)acetophenone
Formula:
C₁₄H₁₀BrFO₂ |
MDL Number:
MFCD06411656 CAS Number: 845866-51-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...