4'-(3-Bromophenoxy)acetophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

023239-5G 023239-25G
FLUO023239-5GEA 0 GBP
FLUO023239-5G FLUO023239-25G
4'-(3-Bromophenoxy)acetophenone
4'-(3-Bromophenoxy)acetophenone
Formula: C₁₄H₁₁BrO₂
MDL Number: MFCD06411651
CAS Number: 845866-46-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR