3-Butoxy-5-formylphenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (3-Butoxy-5-formylphenyl)boronic acid, 3-butoxy-5-formylphenylboronic acid

BB-8101-1G BB-8101-250M
COBBBB-8101-1GEA 0 GBP
COBBBB-8101-1G COBBBB-8101-250M
3-Butoxy-5-formylphenylboronic acid
3-Butoxy-5-formylphenylboronic acid
Formula: C₁₁H₁₅BO₄
MW: 222.0500 g/mol
MDL Number: MFCD08705271
CAS Number: 1072951-70-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR