1-Iodo-4-methylimidazolium chloride

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Iodo-1-methyl-1H-imidazole hydrochloride

HI-1155-5G HI-1155-1G
COBBHI-1155-5GEA 0 GBP
COBBHI-1155-5G COBBHI-1155-1G
1-Iodo-4-methylimidazolium chloride
1-Iodo-4-methylimidazolium chloride
Formula: C₄H₆ClIN₂
MDL Number: MFCD07783893
CAS Number: 871333-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR