5-Chloro-2-hydroxyphenylboronic acid ≥97%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 5-Chloro-2-hydroxybenzeneboronic acid

H53075.MD H53075.03
H53075.MDEA 0 GBP
H53075.MD H53075.03
5-Chloro-2-hydroxyphenylboronic acid ≥97%
5-Chloro-2-hydroxyphenylboronic acid

Purity: 97% CAS No.: 89488-25-5 Molecular Formula: C6H6BClO3 Formula Weight: 172.37 Melting Point: 166-170° MDL No.: MFCD06659863

Formula: C₆H₆BClO₃
MW: 172.37 g/mol
Melting Pt: 166…170 °C
Storage Temperature: Ambient
MDL Number: MFCD06659863
CAS Number: 89488-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR