2,6-Dichloro-3-formylphenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-9043-5G BB-9043-1G
COBBBB-9043-5GEA 0 GBP
COBBBB-9043-5G COBBBB-9043-1G
2,6-Dichloro-3-formylphenylboronic acid
2,6-Dichloro-3-formylphenylboronic acid
Formula: C₇H₅BCl₂O₃
MDL Number: MFCD06796322
CAS Number: 1218790-87-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR