3-Chloro-4-hydroxyphenylboronic acid pinacol ester
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
PN-8794-5G
PN-8794-1G
COBBPN-8794-5GEA
0
GBP
COBBPN-8794-5G
COBBPN-8794-1G
3-Chloro-4-hydroxyphenylboronic acid pinacol ester
3-Chloro-4-hydroxyphenylboronic acid pinacol ester
Formula:
C₁₂H₁₆BClO₃ MW: 254.5200 g/mol |
MDL Number:
MFCD09260490 CAS Number: 629658-06-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...