N-tert-Butyl-4-bromo-2-nitroaniline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AN-1493-1G
AN-1493-5G
AN-1493-250M
COBBAN-1493-1GEA
0
GBP
COBBAN-1493-1G
COBBAN-1493-5G
COBBAN-1493-250M
N-tert-Butyl-4-bromo-2-nitroaniline
N-tert-Butyl-4-bromo-2-nitroaniline
Formula:
C₁₀H₁₃BrN₂O₂ |
MDL Number:
MFCD12756414 CAS Number: 1135351-95-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...