N-tert-Butyl-2-bromo-4-nitroaniline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AN-1428-5G
AN-1428-1G
COBBAN-1428-5GEA
0
GBP
COBBAN-1428-5G
COBBAN-1428-1G
N-tert-Butyl-2-bromo-4-nitroaniline
N-tert-Butyl-2-bromo-4-nitroaniline
Formula:
C₁₀H₁₃BrN₂O₂ |
MDL Number:
MFCD11855829 CAS Number: 952664-76-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...