5-Bromo-4-methoxy-2-nitroaniline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AN-2571-5G
AN-2571-1G
AN-2571-250M
COBBAN-2571-5GEA
0
GBP
COBBAN-2571-5G
COBBAN-2571-1G
COBBAN-2571-250M
5-Bromo-4-methoxy-2-nitroaniline
5-Bromo-4-methoxy-2-nitroaniline
Formula:
C₇H₇BrN₂O₃ MW: 247.0461 g/mol |
MDL Number:
MFCD00031127 CAS Number: 6943-69-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...