N-Ethyl-3-boronobenzenesulphonamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-2615-5G BB-2615-1G
COBBBB-2615-5GEA 0 GBP
COBBBB-2615-5G COBBBB-2615-1G
N-Ethyl-3-boronobenzenesulphonamide
N-Ethyl 3-boronobenzenesulphonamide
Formula: C₈H₁₂BNO₄S
MDL Number: MFCD07783855
CAS Number: 871329-76-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR