N,N-Dimethyl 4-boronobenzenesulphonamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-(N,N-Dimethylsulphamoyl)phenylboronic acid, 4-(Dimethylsulphamoyl)benzeneboronic acid

BB-2424-1G BB-2424-5G
COBBBB-2424-1GEA 0 GBP
COBBBB-2424-1G COBBBB-2424-5G
N,N-Dimethyl 4-boronobenzenesulphonamide
N,N-Dimethyl 4-boronobenzenesulphonamide
Formula: C₈H₁₂BNO₄S
MW: 229.06 g/mol
MDL Number: MFCD06659849
CAS Number: 486422-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR