N,N-Diethyl 4-boronobenzenesulphonamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (4-(N,N-Diethylsulphamoyl)phenyl)boronic acid, 4-(Diethylsulphamoyl)benzeneboronic acid

BB-2420-5G BB-2420-1G
COBBBB-2420-5GEA 0 GBP
COBBBB-2420-5G COBBBB-2420-1G
N,N-Diethyl 4-boronobenzenesulphonamide
N,N-Diethyl 4-boronobenzenesulphonamide
Formula: C₈H₁₂BNO₄S
MW: 257.11 g/mol
MDL Number: MFCD06659846
CAS Number: 850568-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR