N,N-Diethyl 2-boronobenzenesulphonamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-N,N-Diethylsulphamoylphenylboronic acid

BB-3875-5G BB-3875-1G
COBBBB-3875-5GEA 0 GBP
COBBBB-3875-5G COBBBB-3875-1G
N,N-Diethyl 2-boronobenzenesulphonamide
N,N-Diethyl 2-boronobenzenesulphonamide
Formula: C₁₀H₁₆BNO₄S
MW: 257.1143 g/mol
MDL Number: MFCD09258634
CAS Number: 957061-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR