Hesperetin, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3',5,7-Trihydroxy-4'-methoxyflavanone, 4'-Methoxy-3',5,7-trihydroxyflavanone, (S)-5,7-Dihydroxy-2-(3-hydroxy-4-methoxyphenyl)chroman-4-one, (±)-Hesperetin
W431300-50G
W431300-5G
SIALW431300-50GEA
508
GBP
SIALW431300-50G
SIALW431300-5G
Hesperetin, Sigma-Aldrich®
Hesperetin
Formula:
C₁₆H₁₄O₆ MW: 302.30 g/mol |
MDL Number:
MFCD00075646 CAS Number: 520-33-2 EINECS: 208-290-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...