Sodium acetate-[¹³C₂] (99% ¹³C ), Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ200

282014-1G 282014-250MG
SIAL282014-1GEA 710 GBP
SIAL282014-1G SIAL282014-250MG
Sodium acetate-[¹³C₂] (99% ¹³C ), Sigma-Aldrich®
Sodium acetate-[13C2]
Formula: H₃CCOONa
Storage Temperature: Ambient
MDL Number: MFCD00037667
CAS Number: 56374-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR