2-Piperazinonicotinonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009836-1G 009836-5G 009836-25G
FLUO009836-1GEA 0 GBP
FLUO009836-1G FLUO009836-5G FLUO009836-25G
2-Piperazinonicotinonitrile
2-Piperazinonicotinonitrile
Formula: C₁₀H₁₂N₄
MDL Number: MFCD00190248
CAS Number: 84951-44-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR