trans,trans-Dibenzylideneacetone

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS23086G100 AS23086G25
ACTIAS23086G100EA 0 GBP
ACTIAS23086G100 ACTIAS23086G25
trans,trans-Dibenzylideneacetone
trans,trans-Dibenzylideneacetone
Formula: C₁₇H₁₄O
MDL Number: MFCD00004790
CAS Number: 35225-79-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR