4-Benzyloxy-2-chloro-5-pyrimidineboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Benzyloxy-2-chloropyrimidine-5-boronic acid pinacol ester

PN-8689-1G PN-8689-250M PN-8689-100M
COBBPN-8689-1GEA 0 GBP
COBBPN-8689-1G COBBPN-8689-250M COBBPN-8689-100M
4-Benzyloxy-2-chloro-5-pyrimidineboronic acid pinacol ester
4-Benzyloxy-2-chloro-5-pyrimidineboronic acid pinacol ester
Formula: C₁₇H₂₀BClN₂O₃
MW: 346.62 g/mol
MDL Number: MFCD06798267
CAS Number: 1073354-22-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR