2,4-Di-tert-butoxy-5-pyrimidineboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(2,4-Di-tert-butoxypyrimidin-5-yl)boronic acid
BB-2238-1G
BB-2238-5G
COBBBB-2238-1GEA
0
GBP
COBBBB-2238-1G
COBBBB-2238-5G
2,4-Di-tert-butoxy-5-pyrimidineboronic acid
2,4-Di-tert-butoxy-5-pyrimidineboronic acid
Formula:
C₁₂H₂₁BN₂O₄ MW: 268.117 g/mol |
MDL Number:
MFCD01318980 CAS Number: 109299-79-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...