2-(Dimethylamino)-5-pyrimidineboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (2-(Dimethylamino)pyrimidin-5-yl)boronic acid, 2-Dimethylaminopyrimidinyl-5-boronic acid

BB-8704-1G BB-8704-5G
COBBBB-8704-1GEA 31.66 GBP
COBBBB-8704-1G COBBBB-8704-5G
2-(Dimethylamino)-5-pyrimidineboronic acid
2-(Dimethylamino)-5-pyrimidineboronic acid
Formula: C₆H₁₀BN₃O₂
MW: 166.9735 g/mol
MDL Number: MFCD10696933
CAS Number: 756817-82-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR