2-Amino-5-pyrimidineboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Amino-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine, 2-Aminopyrimidine-5-boronic acid Pinacol ester, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine, 2-(2-Aminopyrimidin-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

PN-8680-5G PN-8680-1G
COBBPN-8680-5GEA 0 GBP
COBBPN-8680-5G COBBPN-8680-1G
2-Amino-5-pyrimidineboronic acid pinacol ester
2-Amino-5-pyrimidineboronic acid pinacol ester
Formula: C₁₀H₁₆BN₃O₂
MW: 221.06 g/mol
MDL Number: MFCD06795685
CAS Number: 402960-38-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR