2-Amino-5-pyrimidineboronic acid pinacol ester

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Amino-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine, 2-Aminopyrimidine-5-boronic acid Pinacol ester, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine, 2-(2-Aminopyrimidin-5-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

AS22702G1 AS22702G25 AS22702G5
ACTIAS22702G1EA 17 GBP
ACTIAS22702G1 ACTIAS22702G25 ACTIAS22702G5
2-Amino-5-pyrimidineboronic acid pinacol ester
2-Amino-5-pyrimidineboronic acid pinacol ester
Formula: C₁₀H₁₆BN₃O₂
MW: 221.06 g/mol
MDL Number: MFCD06795685
CAS Number: 402960-38-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR