3'-Fluoro-4'-(1-pyrazolyl)acetophenone

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21558G25 AS21558G5
ACTIAS21558G25EA 0 GBP
ACTIAS21558G25 ACTIAS21558G5
3'-Fluoro-4'-(1-pyrazolyl)acetophenone
3'-Fluoro-4'-(1-pyrazolyl)acetophenone
Formula: C₁₁H₉FN₂O
MDL Number: MFCD11136849
CAS Number: 1152964-31-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR