4'-(1-Pyrazolyl)acetophenone
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS21540G25
AS21540G5
ACTIAS21540G25EA
361
GBP
ACTIAS21540G25
ACTIAS21540G5
4'-(1-Pyrazolyl)acetophenone
4'-(1-Pyrazolyl)acetophenone
Formula:
C₁₁H₁₀N₂O MW: 186.2099 g/mol |
MDL Number:
MFCD08572143 CAS Number: 25699-98-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...