5-Amino-3-(4-bromophenyl)-1H-pyrazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(4-Bromophenyl)-1H-pyrazol-5-amine

HC-2151-5G HC-2151-1G
COBBHC-2151-5GEA 0 GBP
COBBHC-2151-5G COBBHC-2151-1G
5-Amino-3-(4-bromophenyl)-1H-pyrazole
5-Amino-3-(4-bromophenyl)-1H-pyrazole
Formula: C₉H₈BrN₃
MDL Number: MFCD00558911
CAS Number: 78583-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR