3'-Bromo-4'-(1-pyrazolyl)acetophenone

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS21561G25 AS21561G5
ACTIAS21561G25EA 0 GBP
ACTIAS21561G25 ACTIAS21561G5
3'-Bromo-4'-(1-pyrazolyl)acetophenone
3'-Bromo-4'-(1-pyrazolyl)acetophenone
Formula: C₁₁H₉BrN₂O
MDL Number: MFCD11656810
CAS Number: 1186663-58-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR