4-Bromo-3,5-dimethyl-1H-pyrazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Bromo-3,5-dimethylpyrazole

AS23177G100 AS23177G25
ACTIAS23177G100EA 0 GBP
ACTIAS23177G100 ACTIAS23177G25
4-Bromo-3,5-dimethyl-1H-pyrazole
4-Bromo-3,5-dimethyl-1H-pyrazole
Formula: C₅H₇BrN₂
MDL Number: MFCD00005242
CAS Number: 3398-16-1
EINECS: 222-263-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR